Methyl 5-amino-6-bromopicolinate 97%

Supplier: Ambeed
Warning

A230578-100MG A230578-250MG
77140-056EA 65.92 USD
77140-056 77140-058
Methyl 5-amino-6-bromopicolinate 97%
Methyl 5-amino-6-bromopicolinate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MDL Number: MFCD16036645
CAS Number: 797060-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR