2-Bromo-5-fluoro-4-nitropyridine 95%

Supplier: Ambeed
Warning

A229691-100MG A229691-250MG A229691-5G A229691-1G
77140-042EA 24.73 USD
77140-042 77140-044 77568-222 77746-340
2-Bromo-5-fluoro-4-nitropyridine 95%
2-Bromo-5-fluoro-4-nitropyridine

Precaución: For research use only.

Formula: C₅H₂BrFN₂O₂
MDL Number: MFCD28740491
CAS Number: 1805578-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR