2-Fluoro-4-hydroxybenzonitrile 98%

Supplier: Ambeed
Danger

A229350-10G A229350-25G A229350-100G
77140-040EA 10.3 USD
77140-040 77615-910 77685-625
2-Fluoro-4-hydroxybenzonitrile 98%
2-Fluoro-4-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄FNO
MDL Number: MFCD00192177
CAS Number: 82380-18-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR