2-Bromo-1-fluoro-3-iodobenzene 98%

Supplier: Ambeed
Warning

A229310-1G A229310-250MG A229310-5G
77140-034EA 33.99 USD
77140-034 77140-036 77140-038
2-Bromo-1-fluoro-3-iodobenzene 98%
2-Bromo-1-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
MDL Number: MFCD09056796
CAS Number: 851368-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR