5-Bromo-2-methoxypyridine-3-carboxaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-2-methoxynicotinaldehyde
A228768-10G
A228768-1G
A228768-250MG
A228768-25G
A228768-5G
77140-024EA
253.56
USD
77140-024
77140-026
77140-028
77140-030
77140-032
5-Bromo-2-methoxypyridine-3-carboxaldehyde 97%
5-Bromo-2-methoxypyridine-3-carboxaldehyde
Precaución: for Research Use Only
Formula:
C₇H₆BrNO₂ MW: 216.034 g/mol |
MDL Number:
MFCD05663521 CAS Number: 103058-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...