1-Methyl-7-nitroisatoic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-7-nitro-3,1-benzoxazine-2,4-dione

A228759-100MG A228759-10MG A228759-1G A228759-1MG A228759-250MG A228759-50MG A228759-5MG
77140-004EA 82.4 USD
77140-004 77140-006 77140-008 77140-010 77140-012 77140-014 77140-016
1-Methyl-7-nitroisatoic anhydride 98%
1-Methyl-7-nitroisatoic anhydride

Precaución: for Research Use Only

Formula: C₉H₆N₂O₅
MDL Number: MFCD18432729
CAS Number: 73043-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR