4-Chloro-o-cresol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Chloro-4-hydroxy-3-methylbenzene, 4-Chloro-2-methylphenol
A226673-100G
A226673-10G
A226673-1G
A226673-25G
A226673-500G
77139-948EA
39.26
USD
77139-948
77139-950
77139-952
77139-954
77139-956
4-Chloro-o-cresol 97%
4-Chloro-o-cresol
Precaución: for Research Use Only
Formula:
C₇H₇ClO MW: 142.58 g/mol |
MDL Number:
MFCD00002321 CAS Number: 1570-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...