1-Methylpyrrolidine-2-thione 95%

Supplier: Ambeed
Warning

A226625-1G A226625-25G A226625-5G
77139-938EA 21.22 USD
77139-938 77139-940 77139-942
1-Methylpyrrolidine-2-thione 95%
1-Methylpyrrolidine-2-thione

Precaución: for Research Use Only

Formula: C₅H₉NS
MW: 115.19 g/mol
MDL Number: MFCD00966332
CAS Number: 10441-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR