5-(tert-Butyl)-1H-indole 95%
Supplier: Ambeed
|
Warning
|
A225567-100MG
A225567-1G
A225567-250MG
77139-928EA
28.84
USD
77139-928
77139-930
77139-932
5-(tert-Butyl)-1H-indole 95%
5-(tert-Butyl)-1H-indole
Precaución: for Research Use Only
Formula:
C₁₂H₁₅N |
MDL Number:
MFCD20668007 CAS Number: 92462-70-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...