Bis-PEG3-acid 95%

Supplier: Ambeed
Warning

Synonyms: 3,3'-((Oxybis(ethane-2,1-diyl))bis(oxy))dipropanoic acid

A224911-1G A224911-5G A224911-100MG A224911-250MG
77139-910EA 166.57 USD
77139-910 77139-912 77746-324 77746-325
Bis-PEG3-acid 95%
Bis-PEG3-acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₇
MW: 250.246 g/mol
MDL Number: MFCD22056316
CAS Number: 96517-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR