4-((3-(2H-1,2,3-Triazol-2-yl)phenyl)amino)-2-(((1R,2S)-2-aminocyclohexyl)amino)pyrimidine-5-carboxamide hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
PRT062607 HCl
A224641-10MG
A224641-1MG
A224641-25MG
A224641-50MG
A224641-5MG
77139-900EA
62.82
USD
77139-900
77139-902
77139-904
77139-906
77139-908
4-((3-(2H-1,2,3-Triazol-2-yl)phenyl)amino)-2-(((1R,2S)-2-aminocyclohexyl)amino)pyrimidine-5-carboxamide hydrochloride 98%
4-((3-(2H-1,2,3-Triazol-2-yl)phenyl)amino)-2-(((1R,2S)-2-aminocyclohexyl)amino)pyrimidine-5-carboxamide hydrochloride
Precaución: for Research Use Only
Formula:
C₁₉H₂₄ClN₉O |
MDL Number:
MFCD25976708 CAS Number: 1370261-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...