(R)-(+)-4-Fluoro-α-methylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(4-Fluorophenyl)ethanol

A223552-100MG A223552-250MG A223552-5G
77139-890EA 9.55 USD
77139-890 77139-892 77139-894
(R)-(+)-4-Fluoro-α-methylbenzyl alcohol 97%
(R)-(+)-4-Fluoro-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD03092997
CAS Number: 101219-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR