3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl acrylate 95%

Supplier: Ambeed
Warning

A223507-10G A223507-1G A223507-25G A223507-5G
77139-882EA 30.9 USD
77139-882 77139-884 77139-886 77139-888
3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl acrylate 95%
3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl acrylate

Precaución: for Research Use Only

Formula: C₁₅H₃₆O₅Si₄
MDL Number: MFCD00054802
CAS Number: 17096-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR