4-Iodo-2-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-1-methoxy-2-nitrobenzene

A222510-1G A222510-250MG A222510-25G A222510-5G
77139-862EA 22.66 USD
77139-862 77139-864 77139-866 77139-868
4-Iodo-2-nitroanisole 98%
4-Iodo-2-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆INO₃
MDL Number: MFCD08060933
CAS Number: 52692-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR