Benzyl (3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 97%

Supplier: Ambeed
Warning

A221375-1G A221375-250MG A221375-5G
77139-848EA 83.82 USD
77139-848 77139-850 77139-852
Benzyl (3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 97%
Benzyl (3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₀H₂₃BFNO₄
MW: 371.21 g/mol
MDL Number: MFCD22192533
CAS Number: 2096997-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR