(S)-(+)-1,2-Propanediol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(+)-1,2-Propylene glykol, (S)-Propane-1,2-diol, (S)-(+)-1,2-Dihydroxy propane
A219631-1000G
A219631-100G
A219631-10G
A219631-25G
A219631-500G
A219631-5G
77139-832EA
160.2
USD
77139-832
77139-834
77139-836
77139-838
77139-840
77139-842
(S)-(+)-1,2-Propanediol 98%
(S)-(+)-1,2-Propanediol
Precaución: for Research Use Only
Formula:
C₃H₈O₂ MW: 76.1 g/mol |
MDL Number:
MFCD00004539 CAS Number: 4254-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...