Allyl 2-((2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)ethyl)amino)acetate hydrochloride 97%

Supplier: Ambeed
Warning

A218014-1G A218014-250MG A218014-25G A218014-5G
77139-802EA 52.53 USD
77139-802 77139-804 77139-806 77139-808
Allyl 2-((2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)ethyl)amino)acetate hydrochloride 97%
Allyl 2-((2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)ethyl)amino)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₅ClN₂O₄
MDL Number: MFCD28404628
CAS Number: 861114-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR