1,1'-Binaphthyl-2,2'-diamine 97%

Supplier: Ambeed
Warning

A217671-100G A217671-10G A217671-1G A217671-250MG A217671-25G A217671-5G
77139-790EA 221.45 USD
77139-790 77139-792 77139-794 77139-796 77139-798 77139-800
1,1'-Binaphthyl-2,2'-diamine 97%
1,1'-Binaphthyl-2,2'-diamine

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂
MDL Number: MFCD00145204
CAS Number: 4488-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR