(3-Chloro-4-fluoro-5-isopropoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-fluoro-5-isopropoxyphenylboronic acid

A217286-1G A217286-250MG A217286-5G
77139-778EA 133.68 USD
77139-778 77139-780 77139-782
(3-Chloro-4-fluoro-5-isopropoxyphenyl)boronic acid 97%
(3-Chloro-4-fluoro-5-isopropoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BClFO₃
MW: 232.44 g/mol
MDL Number: MFCD20441835
CAS Number: 2096331-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR