(1H-Pyrazol-3-yl)methanol 96%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)pyrazole

A216182-10G A216182-1G A216182-250MG A216182-5G
77139-748EA 203.95 USD
77139-748 77139-750 77139-752 77139-754
(1H-Pyrazol-3-yl)methanol 96%
(1H-Pyrazol-3-yl)methanol

Precaución: for Research Use Only

Formula: C₄H₆N₂O
MW: 98.103 g/mol
MDL Number: MFCD08436148
CAS Number: 23585-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR