2,4-Difluorobenzoic acid 98%

Supplier: Ambeed
Warning

A215980-1000G A215980-100G A215980-10G A215980-25G A215980-500G A215980-5G
77139-736EA 206.88 USD
77139-736 77139-738 77139-740 77139-742 77139-744 77139-746
2,4-Difluorobenzoic acid 98%
2,4-Difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
MDL Number: MFCD00011670
CAS Number: 1583-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR