(S,E)-3-(2,6-Dichloro-4-((4-(3-(1-(hexyloxy)ethyl)-2-methoxyphenyl)thiazol-2-yl)carbamoyl)phenyl)-2-methylacrylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Lusutrombopag
A215820-10MG
A215820-1MG
A215820-2MG
A215820-5MG
A215820-250MG
77139-730EA
53.05
USD
77139-730
77139-732
77139-734
77615-814
77685-581
(S,E)-3-(2,6-Dichloro-4-((4-(3-(1-(hexyloxy)ethyl)-2-methoxyphenyl)thiazol-2-yl)carbamoyl)phenyl)-2-methylacrylic acid 97%
(S,E)-3-(2,6-Dichloro-4-((4-(3-(1-(hexyloxy)ethyl)-2-methoxyphenyl)thiazol-2-yl)carbamoyl)phenyl)-2-methylacrylic acid
Precaución: for Research Use Only
Formula:
C₂₉H₃₂Cl₂N₂O₅S MW: 591.55 g/mol |
MDL Number:
MFCD28502075 CAS Number: 1110766-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...