3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A215484-1G A215484-250MG
77139-726EA 148.53 USD
77139-726 77139-728
3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
3-(Trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C5H3F3N2O
MW: 164.0900 g/mol
MDL Number: MFCD07375188
CAS Number: 1001020-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR