6-Chloro-2-benzoxazolinone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-1,3-benzoxazol-2(3H)-one, 6-Chlorobenzo[d]oxazol-2(3H)-one, 6-Chloro-2-hydroxybenzoxazole, 6-Chloro-2-benzoxazolone

A214607-100G A214607-1G A214607-25G A214607-5G
77139-706EA 259.93 USD
77139-706 77139-708 77139-710 77139-712
6-Chloro-2-benzoxazolinone 98%
6-Chloro-2-benzoxazolinone

Precaución: for Research Use Only

Formula: C₇H₄ClNO₂
MW: 169.56 g/mol
MDL Number: MFCD00463914
CAS Number: 19932-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR