4-(Benzyloxycarbonylamino)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Benzyloxycarbonylamino)benzeneboronic acid, 4-(Benzyloxycarbonylamino)phenylboronic acid, (4-(((Benzyloxy)carbonyl)amino)phenyl)boronic acid, 4-(Cbz-amino)phenylboronic acid, 4-(Cbz-amino)benzeneboronic acid

A214124-10G A214124-1G A214124-250MG A214124-5G
77139-692EA 153.47 USD
77139-692 77139-694 77139-696 77139-698
4-(Benzyloxycarbonylamino)benzeneboronic acid 98%
4-(Benzyloxycarbonylamino)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄BNO₄
MDL Number: MFCD02093055
CAS Number: 192804-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR