(3,4-Dichloro-2-methoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dichloro-2-methoxyphenylboronic acid

A213969-1G A213969-250MG A213969-5G
77139-686EA 133.68 USD
77139-686 77139-688 77139-690
(3,4-Dichloro-2-methoxyphenyl)boronic acid 97%
(3,4-Dichloro-2-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₃
MW: 220.85 g/mol
MDL Number: MFCD18434427
CAS Number: 2096329-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR