(R)-4-(tert-Butoxy)-2-methyl-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

A213232-1G A213232-250MG
77139-666EA 198.39 USD
77139-666 77139-668
(R)-4-(tert-Butoxy)-2-methyl-4-oxobutanoic acid 98%
(R)-4-(tert-Butoxy)-2-methyl-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
MDL Number: MFCD01862170
CAS Number: 185836-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR