7-Cyclobutyl-3-(2,6-difluorophenyl)-6-((1-methyl-1H-1,2,4-triazol-5-yl)methoxy)-[1,2,4]triazolo[4,3-b]pyridazine 97%

Supplier: Ambeed
Warning

A212464-10MG A212464-1MG A212464-5MG
77139-644EA 380.87 USD
77139-644 77139-646 77139-648
7-Cyclobutyl-3-(2,6-difluorophenyl)-6-((1-methyl-1H-1,2,4-triazol-5-yl)methoxy)-[1,2,4]triazolo[4,3-b]pyridazine 97%
7-Cyclobutyl-3-(2,6-difluorophenyl)-6-((1-methyl-1H-1,2,4-triazol-5-yl)methoxy)-[1,2,4]triazolo[4,3-b]pyridazine

Precaución: for Research Use Only

Formula: C₁₉H₁₇F₂N₇O
MW: 397.38 g/mol
MDL Number: MFCD22683821
CAS Number: 233275-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR