Cabozantinib S-malate 99+%

Supplier: Ambeed
Warning

A212266-10MG A212266-1G A212266-1MG A212266-250MG A212266-5MG A212266-100MG A212266-50MG A212266-5G
77139-622EA 8.23 USD
77139-622 77139-624 77139-626 77139-628 77139-630 77685-558 77685-559 77685-560
Cabozantinib S-malate 99+%
Cabozantinib S-malate

Precaución: for Research Use Only

Formula: C₃₂H₃₀FN₃O₁₀
MDL Number: MFCD20923480
CAS Number: 1140909-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR