(1R,2R)-Fmoc-Acpc 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1R,2R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopentanecarboxylic acid
A212031-100MG
A212031-1G
A212031-250MG
A212031-5G
77139-610EA
95.49
USD
77139-610
77139-612
77139-614
77139-616
(1R,2R)-Fmoc-Acpc 95%
(1R,2R)-Fmoc-Acpc
Precaución: for Research Use Only
Formula:
C₂₁H₂₁NO₄ MW: 351.41 g/mol |
MDL Number:
MFCD04112693 CAS Number: 359586-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...