Bromo-PEG5-alcohol 96%

Supplier: Ambeed
Warning

A210876-100MG A210876-250MG A210876-1G A210876-5G
77139-598EA 39.14 USD
77139-598 77593-376 77615-782 77615-784
Bromo-PEG5-alcohol 96%
Bromo-PEG5-alcohol

Precaución: for Research Use Only

Formula: C₁₀H₂₁BrO₅
MDL Number: MFCD18252202
CAS Number: 957205-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR