(S)-tert-Butyl (4-chloro-3-oxo-1-phenylbutan-2-yl)carbamate 97%

Supplier: Ambeed
Danger

A209237-100G A209237-10G A209237-500G A209237-5G A209237-25G
77139-562EA 109.28 USD
77139-562 77139-564 77139-566 77139-568 77593-362
(S)-tert-Butyl (4-chloro-3-oxo-1-phenylbutan-2-yl)carbamate 97%
(S)-tert-Butyl (4-chloro-3-oxo-1-phenylbutan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₀ClNO₃
MW: 297.77 g/mol
MDL Number: MFCD11041198
CAS Number: 102123-74-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR