2-Iodo-5-nitrotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Iodo-2-methyl-4-nitrobenzene

A209045-100G A209045-25G A209045-5G
77139-556EA 112.46 USD
77139-556 77139-558 77139-560
2-Iodo-5-nitrotoluene 98%
2-Iodo-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆INO₂
MW: 263.03 g/mol
MDL Number: MFCD00051517
CAS Number: 5326-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR