D-Panthenol 99%

Supplier: Ambeed
Warning

A208721-100G A208721-10G
77139-552EA 11.33 USD
77139-552 77139-554
D-Panthenol 99%
D-Panthenol

Precaución: for Research Use Only

Formula: C₉H₁₉NO₄
MDL Number: MFCD00065006
CAS Number: 81-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR