2-Fluoro-5-methylphenyl isocyanate 95%

Supplier: Ambeed
Danger

Synonyms: 1-Fluoro-2-isocyanato-4-methylbenzene, 4-Fluoro-3-isocyanatotoluene, 2-Fluoro-5-methylphenylisocyanate

A206351-1G A206351-250MG
77139-514EA 28.65 USD
77139-514 77139-516
2-Fluoro-5-methylphenyl isocyanate 95%
2-Fluoro-5-methylphenyl isocyanate

Precaución: for Research Use Only

Formula: C₈H₆FNO
MW: 151.14 g/mol
MDL Number: MFCD00673050
CAS Number: 190774-50-6
UN: 2206
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR