XL147 98+%

Supplier: Ambeed
Warning

Synonyms: 2-amino-N-[3-[[[3-[(2-chloro-5-methoxyphenyl)amino]-2-quinoxalinyl]amino]sulphonyl]phenyl]-2-methyl-propanamide, 2-Amino-N-(3-(N-(3-((2-chloro-5-methoxyphenyl)amino)quinoxalin-2-yl)sulphamoyl)phenyl)-2-methylpropanamide, SAR245408, Pilaralisib

A205476-10MG A205476-1MG A205476-50MG A205476-5MG
77139-486EA 25.47 USD
77139-486 77139-488 77139-490 77139-492
XL147 98+%
XL147

Precaución: for Research Use Only

Formula: C₂₅H₂₅ClN₆O₄S
MW: 541.02 g/mol
MDL Number: MFCD26142605
CAS Number: 934526-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR