LOXO-101 98%

Supplier: Ambeed
Warning

Synonyms: ARRY 470, Larotrectinib, (S)-N-(5-((R)-2-(2,5-Difluorophenyl)pyrrolidin-1-yl)pyrazolo[1,5-a]pyrimidin-3-yl)-3-hydroxypyrrolidine-1-carboxamide, (3S)-N-[5-[(2R)-2-(2,5-Difluorophenyl)-1-pyrrolidinyl]pyrazolo[1,5-a]pyrimidin-3-yl]-3-hydroxy-1-pyrrolidinecarboxamide

A205378-10MG A205378-1MG A205378-25MG A205378-50MG A205378-5MG
77139-472EA 26.53 USD
77139-472 77139-474 77139-476 77139-478 77139-480
LOXO-101 98%
LOXO-101

Precaución: for Research Use Only

Formula: C₂₁H₂₂F₂N₆O₂
MW: 428.44 g/mol
MDL Number: MFCD28902192
CAS Number: 1223403-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR