3-Chloro-4-(trifluoromethoxy)aniline 98%

Supplier: Ambeed
Warning

A205272-100G A205272-25G A205272-5G
77139-466EA 93.36 USD
77139-466 77139-468 77139-470
3-Chloro-4-(trifluoromethoxy)aniline 98%
3-Chloro-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD00190126
CAS Number: 64628-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR