4-Carboxy-3-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Borono-2-fluorobenzoic acid, 4-Carboxy-3-fluorobenzeneboronic acid

A203974-10G A203974-1G A203974-25G A203974-5G
77139-430EA 59.74 USD
77139-430 77139-432 77139-434 77139-436
4-Carboxy-3-fluorophenylboronic acid 97%
4-Carboxy-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₄
MDL Number: MFCD01114671
CAS Number: 120153-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR