5-Methyl-2-propoxypyridine-3-boronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(5-Methyl-2-propoxypyridin-3-yl)boronic acid
A203889-1G
A203889-250MG
A203889-5G
77139-424EA
90.18
USD
77139-424
77139-426
77139-428
5-Methyl-2-propoxypyridine-3-boronic acid 97%
5-Methyl-2-propoxypyridine-3-boronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₄BNO₃ MW: 195.023 g/mol |
MDL Number:
MFCD18783119 CAS Number: 2096332-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...