4-Fluoro-2,5-dimethylbenzoic acid 97%

Supplier: Ambeed
Warning

A203875-100MG A203875-1G A203875-250MG A203875-5G
77139-412EA 140.04 USD
77139-412 77139-414 77139-416 77139-418
4-Fluoro-2,5-dimethylbenzoic acid 97%
4-Fluoro-2,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.16 g/mol
MDL Number: MFCD23712109
CAS Number: 1427082-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR