(6-Methyl-3-pyridinyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-3-pyridinemethanol, 5-Hydroxymethyl-2-methylpyridine, (6-Methylpyridin-3-yl)methanol, 5-Hydroxymethyl-2-picoline

A203221-10G A203221-250MG A203221-25G A203221-5G A203221-1G
77139-390EA 46.35 USD
77139-390 77139-392 77139-394 77139-396 77746-264
(6-Methyl-3-pyridinyl)methanol 97%
(6-Methyl-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.16 g/mol
MDL Number: MFCD08692003
CAS Number: 34107-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR