2-(4-((5,6-Diphenylpyrazin-2-yl)(isopropyl)amino)butoxy)-N-(methylsulfonyl)acetamide 98%

Supplier: Ambeed
Warning

Synonyms: Selexipag

A203007-10MG A203007-1MG A203007-2MG A203007-50MG A203007-5MG
77139-376EA 42.44 USD
77139-376 77139-378 77139-380 77139-382 77139-384
2-(4-((5,6-Diphenylpyrazin-2-yl)(isopropyl)amino)butoxy)-N-(methylsulfonyl)acetamide 98%
2-(4-((5,6-Diphenylpyrazin-2-yl)(isopropyl)amino)butoxy)-N-(methylsulfonyl)acetamide

Precaución: for Research Use Only

Formula: C₂₆H₃₂N₄O₄S
MW: 496.62 g/mol
MDL Number: MFCD10567093
CAS Number: 475086-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR