3-Iodo-2-methyl-1H-indole, N-BOC protected 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-iodo-2-methyl-1H-indole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-iodo-2-methyl-1H-indole

A202696-100MG A202696-250MG A202696-5G
77139-370EA 35.01 USD
77139-370 77139-372 77139-374
3-Iodo-2-methyl-1H-indole, N-BOC protected 97%
3-Iodo-2-methyl-1H-indole, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₄H₁₆INO₂
MW: 357.187 g/mol
MDL Number: MFCD05864787
CAS Number: 877996-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR