trans-4-((2-(Butylamino)-5-(5-(morpholinomethyl)pyridin-2-yl)pyrimidin-4-yl)amino)cyclohexanol 98+%

Supplier: Ambeed
Warning

Synonyms: UNC2250

A202271-10MG A202271-1MG A202271-50MG A202271-5MG
77139-356EA 62.6 USD
77139-356 77139-358 77139-360 77139-362
trans-4-((2-(Butylamino)-5-(5-(morpholinomethyl)pyridin-2-yl)pyrimidin-4-yl)amino)cyclohexanol 98+%
trans-4-((2-(Butylamino)-5-(5-(morpholinomethyl)pyridin-2-yl)pyrimidin-4-yl)amino)cyclohexanol

Precaución: for Research Use Only

Formula: C₂₄H₃₆N₆O₂
MW: 440.58 g/mol
MDL Number: MFCD27992060
CAS Number: 1493694-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR