(3S,4S)-Pyrrolidine-3,4-diol 97%
Supplier: Ambeed
|
Warning
|
A200816-100MG
A200816-1G
A200816-250MG
77139-332EA
67.9
USD
77139-332
77139-334
77139-336
(3S,4S)-Pyrrolidine-3,4-diol 97%
(3S,4S)-Pyrrolidine-3,4-diol
Precaución: for Research Use Only
Formula:
C₄H₉NO₂ MW: 103.11 g/mol |
MDL Number:
MFCD08686737 CAS Number: 90481-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...