(S,S)-(+)-3,6-Octanediol 96%

Supplier: Ambeed
Warning

Synonyms: (3S,6S)-Octane-3,6-diol, (S,S)-(+)-3,6-Dihydroxy octane, (3S,6S)-(+)-3,6-Octanediol

A199098-100MG A199098-1G A199098-250MG
77139-282EA 12.36 USD
77139-282 77139-284 77139-286
(S,S)-(+)-3,6-Octanediol 96%
(S,S)-(+)-3,6-Octanediol

Precaución: for Research Use Only

Formula: C₈H₁₈O₂
MDL Number: MFCD01631088
CAS Number: 136705-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR