2-(2-Phenyl-1H-indol-3-yl)acetic acid 98%

Supplier: Ambeed
Warning

A198576-100MG A198576-1G
77139-274EA 57.67 USD
77139-274 77139-276
2-(2-Phenyl-1H-indol-3-yl)acetic acid 98%
2-(2-Phenyl-1H-indol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₂
MDL Number: MFCD00798608
CAS Number: 4662-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR