3-Iodo-2-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A197815-1G A197815-5G
77139-270EA 19.1 USD
77139-270 77139-272
3-Iodo-2-methylbenzonitrile 98%
3-Iodo-2-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆IN
MW: 243.0444 g/mol
MDL Number: MFCD09025696
CAS Number: 52107-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR