tert-Butyl (4-(hydroxymethyl)thiazol-2-yl)carbamate 98%

Supplier: Ambeed
Warning

A196326-1G A196326-250MG A196326-5G
77139-246EA 35.01 USD
77139-246 77139-248 77139-250
tert-Butyl (4-(hydroxymethyl)thiazol-2-yl)carbamate 98%
tert-Butyl (4-(hydroxymethyl)thiazol-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₃S
MDL Number: MFCD08275703
CAS Number: 494769-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR